N,N-Bis(2-chloroethyl)prop-2-yn-1-amine 95%

Supplier: Ambeed
Danger

A1535580-1G A1535580-250MG
76668-090EA 933 USD
76668-090 77876-194
N,N-Bis(2-chloroethyl)prop-2-yn-1-amine 95%
N,N-Bis(2-chloroethyl)prop-2-yn-1-amine

Precaución: for Research Use Only

Formula: C₇H₁₁Cl₂N
MDL Number: MFCD20623087
CAS Number: 923-82-0
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR