(4-(2-(Dimethylamino)-2-oxoethyl)phenyl)boronic acid 95+%

Supplier: Ambeed
Warning

A1534454-100MG A1534454-1G A1534454-250MG
76668-062EA 37 USD
76668-062 76668-064 76668-066
(4-(2-(Dimethylamino)-2-oxoethyl)phenyl)boronic acid 95+%
(4-(2-(Dimethylamino)-2-oxoethyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄BNO₃
MW: 207.03 g/mol
CAS Number: 1228181-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR