1-(tert-Butyl) 4-ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1,4-dicarboxylate 95+%

Supplier: Ambeed
Warning

A1533714-100MG A1533714-250MG
76668-036EA 37.14 USD
76668-036 76668-038
1-(tert-Butyl) 4-ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1,4-dicarboxylate 95+%
1-(tert-Butyl) 4-ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1,4-dicarboxylate

Precaución: for Research Use Only

Formula: C₂₂H₃₀BNO₆
MW: 415.28 g/mol
CAS Number: 2828439-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR