3-Chloro-N-ethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide 97%

Supplier: Ambeed
Warning

A1533488-100MG A1533488-1G A1533488-250MG
76668-030EA 37.14 USD
76668-030 76668-032 76668-034
3-Chloro-N-ethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide 97%
3-Chloro-N-ethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

Precaución: for Research Use Only

Formula: C₁₅H₂₁BClNO₃
MW: 309.59 g/mol
CAS Number: 2828439-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR