(3-Fluorophenyl)(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol 97%

Supplier: Ambeed
Warning

A1533425-100MG A1533425-1G A1533425-250MG A1533425-5G
76668-020EA 37.14 USD
76668-020 76668-022 76668-024 76668-026
(3-Fluorophenyl)(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol 97%
(3-Fluorophenyl)(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₉H₂₂BFO₃
MW: 328.18 g/mol
CAS Number: 2828439-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR