2-(Benzyloxy)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)pyridine 95%

Supplier: Ambeed
Warning

A1533421-100MG A1533421-1G A1533421-250MG
76668-014EA 19 USD
76668-014 76668-016 76668-018
2-(Benzyloxy)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)pyridine 95%
2-(Benzyloxy)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₁₉H₂₁BF₃NO₃
MW: 379.18 g/mol
MDL Number: MFCD16996443
CAS Number: 2828439-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR