(4-(2-(2-Methoxyethoxy)ethoxy)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A1532782-100MG A1532782-1G A1532782-250MG
76667-970EA 37.14 USD
76667-970 76667-972 76667-974
(4-(2-(2-Methoxyethoxy)ethoxy)phenyl)boronic acid 97%
(4-(2-(2-Methoxyethoxy)ethoxy)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₇BO₅
MW: 240.0607 g/mol
MDL Number: MFCD16198546
CAS Number: 1333122-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR