tert-Butyldimethyl((4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl)methoxy)silane 95+%

Supplier: Ambeed
Warning

A1531391-100MG A1531391-1G A1531391-250MG
76667-946EA 37.14 USD
76667-946 76667-948 76667-950
tert-Butyldimethyl((4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl)methoxy)silane 95+%
tert-Butyldimethyl((4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl)methoxy)silane

Precaución: for Research Use Only

Formula: C₁₇H₃₁BO₃SSi
MW: 354.38 g/mol
CAS Number: 2828439-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR