(3-((2-(Diethylamino)ethyl)carbamoyl)phenyl)boronic acid 95+%

Supplier: Ambeed
Warning

A1530622-100MG A1530622-1G A1530622-250MG
76667-940EA 40 USD
76667-940 76667-942 76667-944
(3-((2-(Diethylamino)ethyl)carbamoyl)phenyl)boronic acid 95+%
(3-((2-(Diethylamino)ethyl)carbamoyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₁BN₂O₃
MDL Number: MFCD28167421
CAS Number: 957103-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR