6-Chloro-2-methylpyrido[3,2-d]pyrimidin-4-amine 95%
Supplier: Ambeed
|
Warning
|
A1530210-1G
A1530210-250MG
76667-924EA
201.58
USD
76667-924
76667-926
6-Chloro-2-methylpyrido[3,2-d]pyrimidin-4-amine 95%
6-Chloro-2-methylpyrido[3,2-d]pyrimidin-4-amine
Precaución: for Research Use Only
Formula:
C₈H₇ClN₄ MW: 194.62 g/mol |
MDL Number:
MFCD34182442 CAS Number: 2561455-12-5 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...