4,4,5,5-Tetramethyl-2-(4-((tetrahydro-2H-pyran-4-yl)methoxy)phenyl)-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Warning

A1527687-100MG A1527687-1G A1527687-250MG
76667-890EA 36.08 USD
76667-890 76667-892 76667-894
4,4,5,5-Tetramethyl-2-(4-((tetrahydro-2H-pyran-4-yl)methoxy)phenyl)-1,3,2-dioxaborolane 95%
4,4,5,5-Tetramethyl-2-(4-((tetrahydro-2H-pyran-4-yl)methoxy)phenyl)-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₈H₂₇BO₄
MW: 318.2156 g/mol
MDL Number: MFCD31583288
CAS Number: 2828439-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR