(3-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)(piperidin-1-yl)methanone 98+%

Supplier: Ambeed
Warning

A1525964-100MG A1525964-1G A1525964-250MG
76667-878EA 36.08 USD
76667-878 76667-880 76667-882
(3-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)(piperidin-1-yl)methanone 98+%
(3-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)(piperidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₈H₂₅BFNO₃
MW: 333.20 g/mol
CAS Number: 2828439-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR