3,5-Dimethyl-1-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-1H-pyrazole 95%

Supplier: Ambeed
Warning

A1523125-100MG A1523125-1G A1523125-250MG A1523125-5G
76667-864EA 38.2 USD
76667-864 76667-866 76667-868 76667-870
3,5-Dimethyl-1-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-1H-pyrazole 95%
3,5-Dimethyl-1-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₇H₂₃BN₂O₂
MW: 298.18 g/mol
MDL Number: MFCD31735798
CAS Number: 2674178-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR