2-(5-Bromopentyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Warning

A1522343-100MG A1522343-1G A1522343-250MG
76667-846EA 37.14 USD
76667-846 76667-848 76667-850
2-(5-Bromopentyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(5-Bromopentyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₁H₂₂BBrO₂
MW: 277.00 g/mol
MDL Number: MFCD29086856
CAS Number: 148562-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR