tert-Butyl ((1H-pyrazol-1-yl)((2,2,2-trifluoroacetyl)imino)methyl)carbamate 95%

Supplier: Ambeed
Warning

A1521840-1G A1521840-250MG
76667-828EA 114.58 USD
76667-828 76667-830
tert-Butyl ((1H-pyrazol-1-yl)((2,2,2-trifluoroacetyl)imino)methyl)carbamate 95%
tert-Butyl ((1H-pyrazol-1-yl)((2,2,2-trifluoroacetyl)imino)methyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₃F₃N₄O₃
MW: 306.24 g/mol
CAS Number: 2028307-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR