1,3-Dioxoisoindolin-2-yl 2-(3-benzoylphenyl)propanoate 97%

Supplier: Ambeed
Warning

A1521331-1G A1521331-250MG A1521331-5G
76667-816EA 45 USD
76667-816 76667-818 76667-820
1,3-Dioxoisoindolin-2-yl 2-(3-benzoylphenyl)propanoate 97%
1,3-Dioxoisoindolin-2-yl 2-(3-benzoylphenyl)propanoate

Precaución: for Research Use Only

Formula: C₂₄H₁₇NO₅
CAS Number: 2241846-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR