N-(2-Bromo-5-fluorophenyl)acetamide 98%

Supplier: Ambeed
Warning

A1519939-25G A1519939-5G
76667-800EA 51 USD
76667-800 77876-050
N-(2-Bromo-5-fluorophenyl)acetamide 98%
N-(2-Bromo-5-fluorophenyl)acetamide

Precaución: for Research Use Only

Formula: C₈H₇BrFNO
MDL Number: MFCD09753710
CAS Number: 1009-06-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR