(S)-2-(Benzo[b]thiophen-2-yl)-4-cyclohexyl-4,5-dihydrooxazole 97%

Supplier: Ambeed
Warning

A1519927-100MG A1519927-1G A1519927-250MG
76667-794EA 150.65 USD
76667-794 77625-172 77625-174
(S)-2-(Benzo[b]thiophen-2-yl)-4-cyclohexyl-4,5-dihydrooxazole 97%
(S)-2-(Benzo[b]thiophen-2-yl)-4-cyclohexyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₇H₁₉NOS
MW: 285.40 g/mol
CAS Number: 2828438-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR