(S)-2-(6-Cyclopropylpyridin-2-yl)-4-methyl-4,5-dihydrooxazole 97%

Supplier: Ambeed
Warning

A1519924-100MG A1519924-1G A1519924-250MG
76667-788EA 178.24 USD
76667-788 77625-160 77625-162
(S)-2-(6-Cyclopropylpyridin-2-yl)-4-methyl-4,5-dihydrooxazole 97%
(S)-2-(6-Cyclopropylpyridin-2-yl)-4-methyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O
MW: 202.2524 g/mol
CAS Number: 2828438-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR