(S)-4-Cyclohexyl-2-(6-cyclopropylpyridin-2-yl)-4,5-dihydrooxazole 97%

Supplier: Ambeed
Warning

A1519880-100MG A1519880-1G A1519880-250MG
76667-726EA 79 USD
76667-726 77625-056 77625-058
(S)-4-Cyclohexyl-2-(6-cyclopropylpyridin-2-yl)-4,5-dihydrooxazole 97%
(S)-4-Cyclohexyl-2-(6-cyclopropylpyridin-2-yl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₇H₂₂N₂O
CAS Number: 2828438-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR