(R)-4-Cyclohexyl-2-(6-cyclopropylpyridin-2-yl)-4,5-dihydrooxazole 97%

Supplier: Ambeed
Warning

A1519879-100MG A1519879-250MG A1519879-1G
76667-724EA 79 USD
76667-724 77625-054 77876-043
(R)-4-Cyclohexyl-2-(6-cyclopropylpyridin-2-yl)-4,5-dihydrooxazole 97%
(R)-4-Cyclohexyl-2-(6-cyclopropylpyridin-2-yl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₇H₂₂N₂O
CAS Number: 2828438-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR