(4R,5S)-2-(6-Cyclopropylpyridin-2-yl)-4,5-diphenyl-4,5-dihydrooxazole 97%
Supplier: Ambeed
|
Warning
|
A1519877-100MG
A1519877-1G
A1519877-250MG
76667-722EA
119.89
USD
76667-722
77625-048
77625-050
(4R,5S)-2-(6-Cyclopropylpyridin-2-yl)-4,5-diphenyl-4,5-dihydrooxazole 97%
(4R,5S)-2-(6-Cyclopropylpyridin-2-yl)-4,5-diphenyl-4,5-dihydrooxazole
Precaución: for Research Use Only
Formula:
C₂₃H₂₀N₂O MW: 340.41 g/mol |
CAS Number:
2828438-62-4 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...