(S)-2-Phenyl-6-(trifluoromethyl)-2,3-dihydroimidazo[1,2-a]pyridine 97%

Supplier: Ambeed
Warning

A1519852-100MG A1519852-1G A1519852-250MG
76667-678EA 57 USD
76667-678 77876-032 77876-033
(S)-2-Phenyl-6-(trifluoromethyl)-2,3-dihydroimidazo[1,2-a]pyridine 97%
(S)-2-Phenyl-6-(trifluoromethyl)-2,3-dihydroimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₁₄H₁₁F₃N₂
MW: 264.24 g/mol
MDL Number: MFCD24848762
CAS Number: 1208867-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR