(R)-4-(tert-Butyl)-2-(6-cyclopropylpyridin-2-yl)-4,5-dihydrooxazole 97%

Supplier: Ambeed
Warning

A1519821-100MG A1519821-250MG
76667-674EA 180.36 USD
76667-674 77624-956
(R)-4-(tert-Butyl)-2-(6-cyclopropylpyridin-2-yl)-4,5-dihydrooxazole 97%
(R)-4-(tert-Butyl)-2-(6-cyclopropylpyridin-2-yl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₅H₂₀N₂O
MW: 244.3321 g/mol
CAS Number: 2828438-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR