(R)-2-(6-Cyclopropylpyridin-2-yl)-4-isopropyl-4,5-dihydrooxazole 97%

Supplier: Ambeed
Warning

A1519819-100MG A1519819-1G A1519819-250MG
76667-670EA 112 USD
76667-670 77624-948 77624-950
(R)-2-(6-Cyclopropylpyridin-2-yl)-4-isopropyl-4,5-dihydrooxazole 97%
(R)-2-(6-Cyclopropylpyridin-2-yl)-4-isopropyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₂O
CAS Number: 2828438-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR