(SP-5-13)-(Acetato-κO)[[2,2'-[(1S,2S)-1,2-cyclohexanediylbis[(nitrilo-κN)methylidyne]]bis[4,6-bis(1,1-dimethylethyl)phenolato-κO]](2-)]cobalt 97%

Supplier: Ambeed
Warning

A1519320-1G A1519320-250MG A1519320-5G
76667-632EA 87 USD
76667-632 76667-634 76667-636
(SP-5-13)-(Acetato-κO)[[2,2'-[(1S,2S)-1,2-cyclohexanediylbis[(nitrilo-κN)methylidyne]]bis[4,6-bis(1,1-dimethylethyl)phenolato-κO]](2-)]cobalt 97%
(SP-5-13)-(Acetato-κO)[[2,2'-[(1S,2S)-1,2-cyclohexanediylbis[(nitrilo-κN)methylidyne]]bis[4,6-bis(1,1-dimethylethyl)phenolato-κO]](2-)]cobalt

Precaución: for Research Use Only

Formula: C₃₈H₅₅CoN₂O₄
MW: 662.78 g/mol
CAS Number: 211821-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR