Benzyl (4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate 98%

Supplier: Ambeed
Warning

A1518261-1G A1518261-250MG A1518261-5G
76667-620EA 217 USD
76667-620 76667-622 76667-624
Benzyl (4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate 98%
Benzyl (4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate

Precaución: for Research Use Only

Formula: C₂₀H₂₃BFNO₄
MW: 371.21 g/mol
MDL Number: MFCD31726487
CAS Number: 2377610-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR