(4-(Cyclopropylmethoxy)-2,6-difluorophenyl)boronic acid 98%

Supplier: Ambeed
Warning

A1518258-1G A1518258-250MG A1518258-5G
76667-614EA 209 USD
76667-614 76667-616 76667-618
(4-(Cyclopropylmethoxy)-2,6-difluorophenyl)boronic acid 98%
(4-(Cyclopropylmethoxy)-2,6-difluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BF₂O₃
MW: 228 g/mol
MDL Number: MFCD30834357
CAS Number: 2377605-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR