3-Formyl-4-nitrobenzoic acid 95%

Supplier: Ambeed
Warning

A1518038-250MG A1518038-1G
76667-588EA 60.48 USD
76667-588 77624-882
3-Formyl-4-nitrobenzoic acid 95%
3-Formyl-4-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅NO₅
MW: 195.1290 g/mol
MDL Number: MFCD22200133
CAS Number: 1092931-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR