3-(3-Hydroxy-phenyl)-acrylic acid ethyl ester 95%

Supplier: Ambeed
Warning

A1517990-250MG A1517990-1G
76667-586EA 34 USD
76667-586 77876-017
3-(3-Hydroxy-phenyl)-acrylic acid ethyl ester 95%
3-(3-Hydroxy-phenyl)-acrylic acid ethyl ester

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₃
MDL Number: MFCD00184677
CAS Number: 96251-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR