trans-Methyl 4-(benzyloxy)cyclohexanecarboxylate 95%

Supplier: Ambeed
Warning

A1517177-100MG A1517177-250MG
76667-582EA 109 USD
76667-582 76667-584
trans-Methyl 4-(benzyloxy)cyclohexanecarboxylate 95%
trans-Methyl 4-(benzyloxy)cyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₀O₃
CAS Number: 1417317-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR