6,6'-((1E,1'E)-(((1R,2R)-Cyclohexane-1,2-diyl)bis(azanylylidene))bis(methanylylidene))bis(4-(tert-butyl)-2-methylphenol) 97%

Supplier: Ambeed
Warning

A1517158-250MG A1517158-1G
76667-572EA 32 USD
76667-572 77876-009
6,6'-((1E,1'E)-(((1R,2R)-Cyclohexane-1,2-diyl)bis(azanylylidene))bis(methanylylidene))bis(4-(tert-butyl)-2-methylphenol) 97%
6,6'-((1E,1'E)-(((1R,2R)-Cyclohexane-1,2-diyl)bis(azanylylidene))bis(methanylylidene))bis(4-(tert-butyl)-2-methylphenol)

Precaución: for Research Use Only

Formula: C₃₀H₄₂N₂O₂
MW: 462.66 g/mol
CAS Number: 1095419-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR