1-(3,5-Bis(trifluoromethyl)phenyl)-3-((S)-1-((R)-2-(4-chlorophenyl)-2-methylpyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)thiourea 95%

Supplier: Ambeed
Danger

A1517156-100MG A1517156-50MG
76667-564EA 220.67 USD
76667-564 76667-566
1-(3,5-Bis(trifluoromethyl)phenyl)-3-((S)-1-((R)-2-(4-chlorophenyl)-2-methylpyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)thiourea 95%
1-(3,5-Bis(trifluoromethyl)phenyl)-3-((S)-1-((R)-2-(4-chlorophenyl)-2-methylpyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)thiourea

Precaución: for Research Use Only

Formula: C₂₆H₂₈ClF₆N₃OS
MW: 580.02 g/mol
CAS Number: 2756689-44-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR