(4S,5S)-1,3-Bis(2-(methoxymethyl)phenyl)-4,5-diphenyl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate 97%

Supplier: Ambeed
Danger

A1517152-100MG A1517152-250MG
76667-560EA 182 USD
76667-560 77876-004
(4S,5S)-1,3-Bis(2-(methoxymethyl)phenyl)-4,5-diphenyl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate 97%
(4S,5S)-1,3-Bis(2-(methoxymethyl)phenyl)-4,5-diphenyl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate

Precaución: for Research Use Only

CAS Number: 1033618-53-9
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR