(4S,5S)-1,3-Bis(2,6-diethylphenyl)-4,5-diphenyl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate 97%

Supplier: Ambeed
Danger

A1517147-100MG A1517147-250MG
76667-558EA 190.97 USD
76667-558 77624-862
(4S,5S)-1,3-Bis(2,6-diethylphenyl)-4,5-diphenyl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate 97%
(4S,5S)-1,3-Bis(2,6-diethylphenyl)-4,5-diphenyl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate

Precaución: for Research Use Only

CAS Number: 1033618-45-9
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR