rel-6-(tert-Butyl) 7a-methyl (4aS,7aS)-tetrahydropyrano[2,3-c]pyrrole-6,7a(2H,7H)-dicarboxylate 97%

Supplier: Ambeed
Warning

A1516871-100MG A1516871-250MG
76667-554EA 127.31 USD
76667-554 76667-556
rel-6-(tert-Butyl) 7a-methyl (4aS,7aS)-tetrahydropyrano[2,3-c]pyrrole-6,7a(2H,7H)-dicarboxylate 97%
rel-6-(tert-Butyl) 7a-methyl (4aS,7aS)-tetrahydropyrano[2,3-c]pyrrole-6,7a(2H,7H)-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₃NO₅
MW: 285.3361 g/mol
CAS Number: 2828438-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR