[[2,2'-[(1R,2R)-1,2-Cyclohexanediylbis[(nitrilo-κN)methylidyne]]bis[4-bis(1,1-dimethylethyl)-6-methyl-phenolato-κO]](2-)](4-methylbenzenesulfonato-κO)cobalt 97%

Supplier: Ambeed
Warning

A1516768-100MG A1516768-1G A1516768-250MG
76667-546EA 77.45 USD
76667-546 76667-548 76667-550
[[2,2'-[(1R,2R)-1,2-Cyclohexanediylbis[(nitrilo-κN)methylidyne]]bis[4-bis(1,1-dimethylethyl)-6-methyl-phenolato-κO]](2-)](4-methylbenzenesulfonato-κO)cobalt 97%
[[2,2'-[(1R,2R)-1,2-Cyclohexanediylbis[(nitrilo-κN)methylidyne]]bis[4-bis(1,1-dimethylethyl)-6-methyl-phenolato-κO]](2-)](4-methylbenzenesulfonato-κO)cobalt

Precaución: for Research Use Only

Formula: C₃₇H₄₇CoN₂O₅S
MW: 690.77 g/mol
CAS Number: 2828438-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR