[[2,2'-[(1S,2S)-1,2-Cyclohexanediylbis[(nitrilo-κN)methylidyne]]bis[4-bis(1,1-dimethylethyl)phenolato-κO]](2-)](4-methylbenzenesulfonato-κO)cobalt 97%

Supplier: Ambeed
Warning

A1516765-100MG A1516765-1G A1516765-250MG
76667-528EA 76.39 USD
76667-528 76667-530 76667-532
[[2,2'-[(1S,2S)-1,2-Cyclohexanediylbis[(nitrilo-κN)methylidyne]]bis[4-bis(1,1-dimethylethyl)phenolato-κO]](2-)](4-methylbenzenesulfonato-κO)cobalt 97%
[[2,2'-[(1S,2S)-1,2-Cyclohexanediylbis[(nitrilo-κN)methylidyne]]bis[4-bis(1,1-dimethylethyl)phenolato-κO]](2-)](4-methylbenzenesulfonato-κO)cobalt

Precaución: for Research Use Only

CAS Number: 2828438-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR