[[2,2'-[(1S,2S)-1,2-Cyclohexanediylbis[(nitrilo-κN)methylidyne]]bis[4-(1,1-dimethylethyl)phenolato-κO]](2-)](nitrato-κO)cobalt 97%
Supplier: Ambeed
|
Danger
|
A1516751-100MG
A1516751-250MG
A1516751-1G
76667-512EA
82.76
USD
76667-512
77597-882
77624-854
[[2,2'-[(1S,2S)-1,2-Cyclohexanediylbis[(nitrilo-κN)methylidyne]]bis[4-(1,1-dimethylethyl)phenolato-κO]](2-)](nitrato-κO)cobalt 97%
[[2,2'-[(1S,2S)-1,2-Cyclohexanediylbis[(nitrilo-κN)methylidyne]]bis[4-(1,1-dimethylethyl)phenolato-κO]](2-)](nitrato-κO)cobalt
Precaución: for Research Use Only
CAS Number:
2828438-27-1 UN: 1479 ADR: 5.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...