3-Methyl-3,5,6,7-tetrahydro-2H-indeno[5,6-b]furan 95%

Supplier: Ambeed
Warning

A1516644-100MG A1516644-250MG
76667-502EA 114 USD
76667-502 76667-504
3-Methyl-3,5,6,7-tetrahydro-2H-indeno[5,6-b]furan 95%
3-Methyl-3,5,6,7-tetrahydro-2H-indeno[5,6-b]furan

Precaución: for Research Use Only

Formula: C₁₂H₁₄O
MW: 174.23 g/mol
CAS Number: 2676863-81-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR