rel-Methyl (3aS,6aR)-5-benzyltetrahydro-1H-thieno[3,4-c]pyrrole-3a(3H)-carboxylate 2,2-dioxide 97%
Supplier: Ambeed
|
Warning
|
A1516409-100MG
A1516409-250MG
76667-494EA
44
USD
76667-494
76667-496
rel-Methyl (3aS,6aR)-5-benzyltetrahydro-1H-thieno[3,4-c]pyrrole-3a(3H)-carboxylate 2,2-dioxide 97%
rel-Methyl (3aS,6aR)-5-benzyltetrahydro-1H-thieno[3,4-c]pyrrole-3a(3H)-carboxylate 2,2-dioxide
Precaución: for Research Use Only
Formula:
C₁₅H₁₉NO₄S |
CAS Number:
2828438-23-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...