4-(2,6-Di(pyrazin-2-yl)pyridin-4-yl)phenol 98%

Supplier: Ambeed
Warning

A1516222-100MG A1516222-1G A1516222-250MG A1516222-5G
76667-482EA 19 USD
76667-482 76667-484 76667-486 77875-990
4-(2,6-Di(pyrazin-2-yl)pyridin-4-yl)phenol 98%
4-(2,6-Di(pyrazin-2-yl)pyridin-4-yl)phenol

Precaución: for Research Use Only

Formula: C₁₉H₁₃N₅O
CAS Number: 1319248-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR