1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclopropanamine hydrochloride 95%

Supplier: Ambeed
Warning

A1515866-100MG A1515866-1G A1515866-250MG
76667-458EA 95.49 USD
76667-458 76667-460 76667-462
1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclopropanamine hydrochloride 95%
1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclopropanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₅H₂₃BClNO₂
MW: 295.6126 g/mol
CAS Number: 2828438-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR