(S)-4-(Pyrrolidin-3-yl)thiomorpholine dihydrochloride 95%

Supplier: Ambeed
Warning

A1515854-100MG A1515854-1G A1515854-250MG
76667-452EA 43 USD
76667-452 76667-454 76667-456
(S)-4-(Pyrrolidin-3-yl)thiomorpholine dihydrochloride 95%
(S)-4-(Pyrrolidin-3-yl)thiomorpholine dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₈Cl₂N₂S
MDL Number: MFCD28023892
CAS Number: 1821826-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR