(R)-4-(Pyrrolidin-3-yl)thiomorpholine dihydrochloride 95%

Supplier: Ambeed
Warning

A1515852-100MG A1515852-1G A1515852-250MG
76667-446EA 36 USD
76667-446 76667-448 76667-450
(R)-4-(Pyrrolidin-3-yl)thiomorpholine dihydrochloride 95%
(R)-4-(Pyrrolidin-3-yl)thiomorpholine dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₈Cl₂N₂S
MW: 245.21 g/mol
MDL Number: MFCD28023893
CAS Number: 1821825-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR