Polyethylene glycol monomethyl ether, M.W. ∼1500
Supplier: Ambeed
|
Warning
|
Synonyms:
Methoxypolyethylene glycols, Methoxy poly(ethylene glycol), PEG MME, Poly(ethylene glycol) methyl ether, mPEG, Methoxypolyethylene glycols-average M.W. 1000, m-PEG-OH
A1515444-500G
A1515444-100G
A1515444-1000G
A1515444-25G
76667-442EA
24
USD
76667-442
77597-866
77624-840
77624-842
Polyethylene glycol monomethyl ether, M.W. ∼1500
Polyethylene glycol monomethyl ether
Precaución: for Research Use Only
Formula:
CH₃(OCH₂CH₂)nOH |
MDL Number:
MFCD00084416 CAS Number: 9004-74-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...