(11bS)-N-(2,6-Bis(4-trifluoromethylphenyl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide 95%
Supplier: Ambeed
|
Warning
|
A1513237-100MG
A1513237-1G
A1513237-250MG
76667-430EA
185.66
USD
76667-430
76667-432
76667-434
(11bS)-N-(2,6-Bis(4-trifluoromethylphenyl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide 95%
(11bS)-N-(2,6-Bis(4-trifluoromethylphenyl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide
Precaución: for Research Use Only
Formula:
C₃₅H₁₉F₉NO₅PS MW: 767.5535 g/mol |
CAS Number:
2757287-55-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...