2,6-Bis(4-methyl-1H-pyrazol-1-yl)pyridine 95%

Supplier: Ambeed
Warning

A1512578-100MG A1512578-1G A1512578-250MG
76667-412EA 39.26 USD
76667-412 76667-414 76667-416
2,6-Bis(4-methyl-1H-pyrazol-1-yl)pyridine 95%
2,6-Bis(4-methyl-1H-pyrazol-1-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₃H₁₃N₅
MW: 239.27 g/mol
CAS Number: 172800-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR