(R)-2-(1,10-Phenanthrolin-2-yl)-4-phenyl-4,5-dihydrooxazole 98%

Supplier: Ambeed
Warning

A1512213-100MG A1512213-250MG
76667-394EA 124.13 USD
76667-394 77597-852
(R)-2-(1,10-Phenanthrolin-2-yl)-4-phenyl-4,5-dihydrooxazole 98%
(R)-2-(1,10-Phenanthrolin-2-yl)-4-phenyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₁H₁₅N₃O
MW: 325.3633 g/mol
CAS Number: 284483-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR