(S)-4-Benzyl-2-(1,10-phenanthrolin-2-yl)-4,5-dihydrooxazole 98%

Supplier: Ambeed
Warning

A1512208-100MG A1512208-250MG A1512208-1G A1512208-5G
76667-388EA 134 USD
76667-388 77624-816 77875-941 77875-942
(S)-4-Benzyl-2-(1,10-phenanthrolin-2-yl)-4,5-dihydrooxazole 98%
(S)-4-Benzyl-2-(1,10-phenanthrolin-2-yl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₂H₁₇N₃O
CAS Number: 2828438-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR